3-Cyclopropylbenzenesulphonyl chloride
Fornitore: Apollo Scientific
|
Pericolo
|
Formula:
C₉H₉ClO₂S Temperatura di stoccaggio: Refrigerata |
MDL Number:
MFCD19201052 CAS Number: 958651-12-2 UN: 3261 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...