Stampa…

Ethyl 5-chlorobenzofuran-2-carboxylate

Fornitore: Apollo Scientific
Attenzione

OR45158-1G OR45158-250MG OR45158-5G
APOSOR45158-1GEA 277 EUR
APOSOR45158-1G APOSOR45158-250MG APOSOR45158-5G
Ethyl 5-chlorobenzofuran-2-carboxylate
Ethyl 5-chlorobenzofuran-2-carboxylate
Formula: C₁₁H₉ClO₃
Peso molecolare: 224,64 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00275273
CAS Number: 59962-89-9

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR