4-Cyclopropyl-benzenemethanol 97%
Fornitore: Apollo Scientific
|
Attenzione
|
Synonyms:
(4-Cyclopropylphenyl)methanol
Formula:
C₁₀H₁₂O Peso molecolare: 148.202 g/mol Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD06802675 CAS Number: 454678-87-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...