Stampa…

Methyl 3-bromo-4-(chlorosulphonyl)benzoate

Fornitore: Apollo Scientific
Pericolo

OR45008-1G OR45008-250MG
APOSOR45008-1GEA 574 EUR
APOSOR45008-1G APOSOR45008-250MG
Methyl 3-bromo-4-(chlorosulphonyl)benzoate
Methyl 3-bromo-4-(chlorosulphonyl)benzoate
Formula: C₈H₆BrClO₄S
Peso molecolare: 313.553 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD20731105
CAS Number: 1354953-47-1
UN: 3261
ADR: 8 III

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR