Methyl 3-ethynylbenzoate 98%
Fornitore: Apollo Scientific
|
Attenzione
|
OR41056-10G
OR41056-1G
OR41056-5G
APOSOR41056-10GEA
317
EUR
APOSOR41056-10G
APOSOR41056-1G
APOSOR41056-5G
Methyl 3-ethynylbenzoate 98%
Methyl 3-ethynylbenzoate
Formula:
C₁₀H₈O₂ Punto di fusione: 45…50 °C Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD08703579 CAS Number: 10602-06-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...