Stampa…

[(Carboxymethoxy)amino](oxo)acetic acid

Fornitore: Apollo Scientific
Pericolo

Synonyms: N-(Carboxymethoxy)oxamic acid

OR40325-100MG OR40325-1G OR40325-500MG
APOSOR40325-100MGEA 279 EUR
APOSOR40325-100MG APOSOR40325-1G APOSOR40325-500MG
[(Carboxymethoxy)amino](oxo)acetic acid
[(Carboxymethoxy)amino](oxo)acetic acid
Formula: C₄H₅NO₆
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD17168268
CAS Number: 177902-90-8

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR