Stampa…

2-Bromo-3-nitrobenzyl bromide

Fornitore: Apollo Scientific
Pericolo

OR400520-1G OR400520-250MG OR400520-5G
APOSOR400520-1GEA 58 EUR
APOSOR400520-1G APOSOR400520-250MG APOSOR400520-5G
2-Bromo-3-nitrobenzyl bromide
2-Bromo-3-nitrobenzyl bromide
Formula: C₇H₅Br₂NO₂
Peso molecolare: 294.928 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD18390263
CAS Number: 82617-49-0
UN: 1759
ADR: 8 III

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR