Stampa…

3-Vinylfuran

Fornitore: Apollo Scientific
Pericolo

OR400372-1G OR400372-250MG
APOSOR400372-1GEA 638 EUR
APOSOR400372-1G APOSOR400372-250MG
3-Vinylfuran
3-Vinylfuran
Formula: C₆H₆O
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD12756842
CAS Number: 67364-02-7
UN: 1993
ADR: 3,III

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR