Stampa…

Methyl 3-bromo-2-hydroxybenzoate

Fornitore: Apollo Scientific

OR400052-1G OR400052-250MG
APOSOR400052-1GEA 97.4 EUR
APOSOR400052-1G APOSOR400052-250MG
Methyl 3-bromo-2-hydroxybenzoate
Methyl 3-bromo-2-hydroxybenzoate
Formula: C₈H₇BrO₃
Peso molecolare: 231,043 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD10566798
CAS Number: 28165-45-9

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR