Stampa…

2-Bromo-5-iodophenylacetonitrile

Fornitore: Apollo Scientific
Pericolo

Synonyms: (2-Bromo-5-iodophenyl)acetonitrile, 2-Bromo-5-iodobenzyl cyanide

OR400041-1G OR400041-250MG OR400041-100MG
APOSOR400041-1GEA 32.1 EUR
APOSOR400041-1G APOSOR400041-250MG APOSOR400041-100MG
2-Bromo-5-iodophenylacetonitrile
2-Bromo-5-iodophenylacetonitrile
Formula: C₈H₅BrIN
Peso molecolare: 321,94 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD18391771
CAS Number: 1261517-16-1
UN: 3439
ADR: 6.1,III

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR