2-(2-Methyl-1,3-dioxolan-2-yl)benzeneboronic acid 97%
Fornitore: Apollo Scientific
Synonyms:
2-(2-Boronobenzene)-2-methyl-1,3-dioxolane
OR3964-1G
OR3964-10G
OR3964-5G
APOSOR3964-1GEA
109
EUR
APOSOR3964-1G
APOSOR3964-10G
APOSOR3964-5G
2-(2-Methyl-1,3-dioxolan-2-yl)benzeneboronic acid 97%
2-(2-Methyl-1,3-dioxolan-2-yl)benzeneboronic acid
Formula:
C₁₀H₁₃BO₄ Peso molecolare: 208,019 g/mol Punto di fusione: 117…120 °C Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD04973737 CAS Number: 243140-14-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...