Stampa…

1,7-Diazaspiro[4.4]nonan-6-one hydrochloride 98%

Fornitore: Apollo Scientific
Attenzione

OR361742-100MG OR361742-250MG
APOSOR361742-100MGEA 172 EUR
APOSOR361742-100MG APOSOR361742-250MG
1,7-Diazaspiro[4.4]nonan-6-one hydrochloride 98%
1,7-Diazaspiro[4.4]nonan-6-one hydrochloride
Formula: C₇H₁₂N₂O·ClH
Peso molecolare: 176,644 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD10700103
CAS Number: 1203682-72-7

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR