Stampa…

4-(Pyrrol-1-yl)benzaldehyde 95+%

Fornitore: Apollo Scientific

Synonyms: 4-(1H-Pyrrol-1-yl)benzaldehyde

OR346717-10G OR346717-5G
APOSOR346717-10GEA 308 EUR
APOSOR346717-10G APOSOR346717-5G
4-(Pyrrol-1-yl)benzaldehyde 95+%
4-(Pyrrol-1-yl)benzaldehyde
Formula: C₁₁H₉NO
Peso molecolare: 171,195 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD03822126
CAS Number: 23351-05-5

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR