Stampa…

1-Methyl-1H-pyrazol-4-ylamine dihydrochloride 95+%

Fornitore: Apollo Scientific

OR346536-1G OR346536-5G
APOSOR346536-1GEA 53.9 EUR
APOSOR346536-1G APOSOR346536-5G
1-Methyl-1H-pyrazol-4-ylamine dihydrochloride 95+%
1-Methyl-1H-pyrazol-4-ylamine dihydrochloride
Formula: C₄H₇N₃·2ClH
Peso molecolare: 170,04 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD09025437
CAS Number: 1063734-49-5,

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR