Stampa…

1-(4-Bromophenyl)-3,5-dimethyl-1H-pyrazole-4-carboxaldehyde 95+%

Fornitore: Apollo Scientific

OR346400-1G OR346400-500MG
APOSOR346400-1GEA 317 EUR
APOSOR346400-1G APOSOR346400-500MG
1-(4-Bromophenyl)-3,5-dimethyl-1H-pyrazole-4-carboxaldehyde 95+%
1-(4-Bromophenyl)-3,5-dimethyl-1H-pyrazole-4-carboxaldehyde
Formula: C₁₂H₁₁BrN₂O
Peso molecolare: 279,133 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD09932059
CAS Number: 294877-39-7

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR