Stampa…

5-Phenyl-[1,2,3]thiadiazole 95+%

Fornitore: Apollo Scientific

OR346390-1G OR346390-5G
APOSOR346390-1GEA 201 EUR
APOSOR346390-1G APOSOR346390-5G
5-Phenyl-[1,2,3]thiadiazole 95+%
5-Phenyl-[1,2,3]thiadiazole
Formula: C₈H₆N₂S
Peso molecolare: 162,212 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00014090
CAS Number: 18212-29-8

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR