Stampa…

Ethyl 5-(4-methoxyphenyl)-[1,2,4]oxadiazole-3-carboxylate

Fornitore: Apollo Scientific

OR346024-1G OR346024-5G
APOSOR346024-1GEA 0 EUR
APOSOR346024-1G APOSOR346024-5G
Ethyl 5-(4-methoxyphenyl)-[1,2,4]oxadiazole-3-carboxylate
Ethyl 5-(4-methoxyphenyl)-[1,2,4]oxadiazole-3-carboxylate
Formula: C₁₂H₁₂N₂O₄
Peso molecolare: 248,235 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD10568207
CAS Number: 151098-14-5

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR