Stampa…

Naphtho[2,1-b]furan-2-carboxylic acid 95%

Fornitore: Apollo Scientific

OR33398-1G OR33398-5G
APOSOR33398-1GEA 328 EUR
APOSOR33398-1G APOSOR33398-5G
Naphtho[2,1-b]furan-2-carboxylic acid 95%
Naphtho[2,1-b]furan-2-carboxylic acid
Formula: C₁₃H₈O₃
Peso molecolare: 212,201 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD01837041
CAS Number: 5656-67-7

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR