Stampa…

2H-Naphtho[2,3-d][1,3]dioxole-2-carboxylic acid 95%

Fornitore: Apollo Scientific

OR33185-1G OR33185-5G
APOSOR33185-1GEA 328 EUR
APOSOR33185-1G APOSOR33185-5G
2H-Naphtho[2,3-d][1,3]dioxole-2-carboxylic acid 95%
2H-Naphtho[2,3-d][1,3]dioxole-2-carboxylic acid
Formula: C₁₂H₈O₄
Peso molecolare: 216,19 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00087796
CAS Number: 200712-25-0

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR