N,N'-bis(4-Methylphenyl)propanediamide 95%
Fornitore: Apollo Scientific
Formula:
C₁₇H₁₈N₂O₂ Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00419232 CAS Number: 5469-94-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...