Stampa…

(9h-Fluoren-9-yl)methyl prop-2-ynylcarbamate 95%

Fornitore: Apollo Scientific

Synonyms: (9H-Fluoren-9-yl)methyl prop-2-yn-1-ylcarbamate

OR323177-1G OR323177-250MG
APOSOR323177-1GEA 85.8 EUR
APOSOR323177-1G APOSOR323177-250MG
(9h-Fluoren-9-yl)methyl prop-2-ynylcarbamate 95%
(9h-Fluoren-9-yl)methyl prop-2-ynylcarbamate
Formula: C₁₈H₁₅NO₂
Peso molecolare: 277,317 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD23704728
CAS Number: 127896-08-6

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR