Stampa…

Methyl 6-methoxybenzo[b]thiophene-2-carboxylate

Fornitore: Apollo Scientific

OR322960-1G OR322960-5G
APOSOR322960-1GEA 357 EUR
APOSOR322960-1G APOSOR322960-5G
Methyl 6-methoxybenzo[b]thiophene-2-carboxylate
Methyl 6-methoxybenzo[b]thiophene-2-carboxylate
Formula: C₁₁H₁₀O₃S
Peso molecolare: 222,26 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD07371543
CAS Number: 550998-58-8

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR