(2-Cyanophenyl)methanesulphonyl chloride
Fornitore: Apollo Scientific
Formula:
C₈H₆ClNO₂S Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD07355846 CAS Number: 51045-34-2 UN: 2923 ADR: 8+6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...