Stampa…

5-(Tetrahydro-2H-pyran-2-yl)thiophene-2-carbaldehyde

Fornitore: Apollo Scientific

OR322573-1G OR322573-250MG
APOSOR322573-1GEA 429 EUR
APOSOR322573-1G APOSOR322573-250MG
5-(Tetrahydro-2H-pyran-2-yl)thiophene-2-carbaldehyde
5-(Tetrahydro-2H-pyran-2-yl)thiophene-2-carbaldehyde
Formula: C₁₀H₁₂O₂S
Peso molecolare: 196,266 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD08059923
CAS Number: 915922-93-9

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR