N-(1-Aminopropan-2-yl)acetamide hydrochloride
Fornitore: Apollo Scientific
|
Attenzione
|
OR321456-1G
OR321456-250MG
APOSOR321456-1GEA
815
EUR
APOSOR321456-1G
APOSOR321456-250MG
N-(1-Aminopropan-2-yl)acetamide hydrochloride
N-(1-Aminopropan-2-yl)acetamide hydrochloride
Formula:
C₅H₁₂N₂O·ClH Peso molecolare: 152,623 g/mol Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD32219462 CAS Number: 1867920-19-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...