Stampa…

(3S,4S)-1-Benzyl-N,4-dimethylpiperidin-3-amine dihydrochloride 97%

Fornitore: Apollo Scientific

OR321455-1G OR321455-5G
APOSOR321455-1GEA 115 EUR
APOSOR321455-1G APOSOR321455-5G
(3S,4S)-1-Benzyl-N,4-dimethylpiperidin-3-amine dihydrochloride 97%
(3S,4S)-1-Benzyl-N,4-dimethylpiperidin-3-amine dihydrochloride
Formula: C₁₄H₂₂N₂·2ClH
Peso molecolare: 291,26 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD11841111
CAS Number: 1354486-07-9

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR