Stampa…

(R)-3-Phenylpiperidine

Fornitore: Apollo Scientific

OR321433-1G OR321433-250MG
APOSOR321433-1GEA 765 EUR
APOSOR321433-1G APOSOR321433-250MG
(R)-3-Phenylpiperidine
(R)-3-Phenylpiperidine
Formula: C₁₁H₁₅N
Peso molecolare: 161,244 g/mol
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD03839934
CAS Number: 430461-56-6

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR