tert-Butyl 3-(cyclopropylamino)-8-azabicyclo[3.2.1]octane-8-carboxylate 95+%
Fornitore: Apollo Scientific
OR318032-1G
OR318032-5G
APOSOR318032-1GEA
442
EUR
APOSOR318032-1G
APOSOR318032-5G
tert-Butyl 3-(cyclopropylamino)-8-azabicyclo[3.2.1]octane-8-carboxylate 95+%
tert-Butyl 3-(cyclopropylamino)-8-azabicyclo[3.2.1]octane-8-carboxylate
Formula:
C₁₅H₂₆N₂O₂ Peso molecolare: 266,379 g/mol Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD13290986 CAS Number: 1305235-96-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...