Stampa…

D-Cyclohexylglycinol

Fornitore: Apollo Scientific

Synonyms: D-CHG-OL

OR316024-1G OR316024-250MG
APOSOR316024-1GEA 181 EUR
APOSOR316024-1G APOSOR316024-250MG
D-Cyclohexylglycinol
D-Cyclohexylglycinol
Formula: C₈H₁₇NO
Peso molecolare: 143,227 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD04112588
CAS Number: 85711-13-3

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR