3,5,6-Trichloro-1,2,4-triazine
Fornitore: Apollo Scientific
OR315527-1G
OR315527-500MG
OR315527-5G
APOSOR315527-1GEA
496
EUR
APOSOR315527-1G
APOSOR315527-500MG
APOSOR315527-5G
3,5,6-Trichloro-1,2,4-triazine
3,5,6-Trichloro-1,2,4-triazine
Formula:
C₃Cl₃N₃ Peso molecolare: 184,411 g/mol Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD09033774 CAS Number: 873-41-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...