Stampa…

(S)-2-Methylpiperidine

Fornitore: Apollo Scientific

OR313089-1G OR313089-5G
APOSOR313089-1GEA 150 EUR
APOSOR313089-1G APOSOR313089-5G
(S)-2-Methylpiperidine
(S)-2-Methylpiperidine
Formula: C₆H₁₃N
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD01862169
CAS Number: 3197-42-0

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR