Stampa…

N-Boc-4-amino-2-butanone

Fornitore: Apollo Scientific

Synonyms: tert-Butyl (3-oxobutyl)carbamate

OR313028-1G OR313028-250MG
APOSOR313028-1GEA 355 EUR
APOSOR313028-1G APOSOR313028-250MG
N-Boc-4-amino-2-butanone
N-Boc-4-amino-2-butanone
Formula: C₉H₁₇NO₃
Peso molecolare: 187,236 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD14539579
CAS Number: 54614-95-8

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR