3,5-Dimethyl-N-(propan-2-yl)aniline
Fornitore: Apollo Scientific
Formula:
C₁₁H₁₇N Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD11145897 CAS Number: 52827-66-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...