Stampa…

1-{[3-(3-Phenyl-1,2,4-oxadiazol-5-yl)pyridin-2-yl]amino}pyrrolidine-2,5-dione

Fornitore: Apollo Scientific

OR311033-1G OR311033-500MG
APOSOR311033-1GEA 539 EUR
APOSOR311033-1G APOSOR311033-500MG
1-{[3-(3-Phenyl-1,2,4-oxadiazol-5-yl)pyridin-2-yl]amino}pyrrolidine-2,5-dione
1-{[3-(3-Phenyl-1,2,4-oxadiazol-5-yl)pyridin-2-yl]amino}pyrrolidine-2,5-dione
Formula: C₁₇H₁₃N₅O₃
Peso molecolare: 335,317 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD14703590
CAS Number: 1232799-90-4

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR