Stampa…

1-(3,4-Dimethylphenyl)-5-(1H-pyrrol-1-yl)-1H-pyrazole-4-carboxylic acid

Fornitore: Apollo Scientific

OR311023-1G OR311023-500MG
APOSOR311023-1GEA 539 EUR
APOSOR311023-1G APOSOR311023-500MG
1-(3,4-Dimethylphenyl)-5-(1H-pyrrol-1-yl)-1H-pyrazole-4-carboxylic acid
1-(3,4-Dimethylphenyl)-5-(1H-pyrrol-1-yl)-1H-pyrazole-4-carboxylic acid
Formula: C₁₆H₁₅N₃O₂
Peso molecolare: 281,309 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD18194582
CAS Number: 1260950-82-0

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR