Stampa…

Ethyl 3-(morpholine-4-sulphonyl)-1H-pyrazole-4-carboxylate

Fornitore: Apollo Scientific

OR311021-1G OR311021-500MG
APOSOR311021-1GEA 539 EUR
APOSOR311021-1G APOSOR311021-500MG
Ethyl 3-(morpholine-4-sulphonyl)-1H-pyrazole-4-carboxylate
Ethyl 3-(morpholine-4-sulphonyl)-1H-pyrazole-4-carboxylate
Formula: C₁₀H₁₅N₃O₅S
Peso molecolare: 289,308 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD18194701
CAS Number: 1108050-18-5

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR