Stampa…

3-[2-(Benzyloxy)phenyl]-5-(chloromethyl)-1,2,4-oxadiazole

Fornitore: Apollo Scientific

OR311002-1G OR311002-500MG
APOSOR311002-1GEA 539 EUR
APOSOR311002-1G APOSOR311002-500MG
3-[2-(Benzyloxy)phenyl]-5-(chloromethyl)-1,2,4-oxadiazole
3-[2-(Benzyloxy)phenyl]-5-(chloromethyl)-1,2,4-oxadiazole
Formula: C₁₆H₁₃ClN₂O₂
Peso molecolare: 300,74 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD09261303
CAS Number: 936497-78-8

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR