Stampa…

1-{3-[3-(3-Chlorophenyl)-1,2,4-oxadiazol-5-yl]pyridin-2-yl}piperidine-4-carboxylic acid

Fornitore: Apollo Scientific

OR311000-1G OR311000-500MG
APOSOR311000-1GEA 539 EUR
APOSOR311000-1G APOSOR311000-500MG
1-{3-[3-(3-Chlorophenyl)-1,2,4-oxadiazol-5-yl]pyridin-2-yl}piperidine-4-carboxylic acid
1-{3-[3-(3-Chlorophenyl)-1,2,4-oxadiazol-5-yl]pyridin-2-yl}piperidine-4-carboxylic acid
Formula: C₁₉H₁₇ClN₄O₃
Peso molecolare: 384,816 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD18201580
CAS Number: 1260934-87-9

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR