3-Ethylmorpholine
Fornitore: Apollo Scientific
|
Attenzione
|
OR308179-1G
OR308179-250MG
APOSOR308179-1GEA
310
EUR
APOSOR308179-1G
APOSOR308179-250MG
3-Ethylmorpholine
3-Ethylmorpholine
Formula:
C₆H₁₃NO Peso molecolare: 115,174 g/mol Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD09054774 CAS Number: 55265-24-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...