Stampa…

(1R,2S)-2-Aminocyclopentanol hydrochloride 97%

Fornitore: Apollo Scientific

OR307677-1G OR307677-250MG
APOSOR307677-1GEA 136 EUR
APOSOR307677-1G APOSOR307677-250MG
(1R,2S)-2-Aminocyclopentanol hydrochloride 97%
(1R,2S)-2-Aminocyclopentanol hydrochloride
Formula: C₅H₁₁NO·ClH
Peso molecolare: 137,608 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD07370091
CAS Number: 137254-03-6

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR