Stampa…

diexo-3-tert-Butoxycarbonylamino-7-oxa-bicyclo[2.2.1]heptane-2-carboxylic acid

Fornitore: Apollo Scientific

OR307562-1G OR307562-250MG
APOSOR307562-1GEA 700 EUR
APOSOR307562-1G APOSOR307562-250MG
diexo-3-tert-Butoxycarbonylamino-7-oxa-bicyclo[2.2.1]heptane-2-carboxylic acid
diexo-3-tert-Butoxycarbonylamino-7-oxa-bicyclo[2.2.1]heptane-2-carboxylic acid
Formula: C₁₂H₁₉NO₅
Peso molecolare: 257,283 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD07783751
CAS Number: 1212286-70-8

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR