Stampa…

2-Amino-3,8-dimethylquinoline hydrochloride

Fornitore: Apollo Scientific

Synonyms: 2-Amino-3,8-dimethyl-1-azanaphthalene hydrochloride, 3,8-Dimethylquinolin-2-amine hydrochloride

OR307518-1G OR307518-5G
APOSOR307518-1GEA 302 EUR
APOSOR307518-1G APOSOR307518-5G
2-Amino-3,8-dimethylquinoline hydrochloride
2-Amino-3,8-dimethylquinoline hydrochloride
Formula: C₁₁H₁₂N₂·ClH
Peso molecolare: 208,687 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD10703337
CAS Number: 1170807-70-1

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR