5-Cyclopropylnicotinic acid
Fornitore: Apollo Scientific
OR306638-1G
OR306638-250MG
APOSOR306638-1GEA
654
EUR
APOSOR306638-1G
APOSOR306638-250MG
5-Cyclopropylnicotinic acid
5-Cyclopropylnicotinic acid
Formula:
C₉H₉NO₂ Punto di fusione: 200…202 °C Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD06802408 CAS Number: 893738-23-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...