Stampa…

4-Iodo-1H-pyrazolo[3,4-b]pyridine

Fornitore: Apollo Scientific

OR305397-1G OR305397-250MG
APOSOR305397-1GEA 232 EUR
APOSOR305397-1G APOSOR305397-250MG
4-Iodo-1H-pyrazolo[3,4-b]pyridine
4-Iodo-1H-pyrazolo[3,4-b]pyridine
Formula: C₆H₄IN₃
Peso molecolare: 245,021 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD11846314
CAS Number: 861881-02-9

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR