1-Oxo-2,3-dihydro-1H-isoindole-5-carboxylic acid
Fornitore: Apollo Scientific
|
Attenzione
|
OR305285-250MG
OR305285-1G
APOSOR305285-250MGEA
757
EUR
APOSOR305285-250MG
APOSOR305285-1G
1-Oxo-2,3-dihydro-1H-isoindole-5-carboxylic acid
1-Oxo-2,3-dihydro-1H-isoindole-5-carboxylic acid
Formula:
C₉H₇NO₃ Peso molecolare: 177,157 g/mol Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD13177824 CAS Number: 23386-40-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...