Stampa…

4-Oxa-1,9-diazaspiro[5.5]undecane dihydrochloride

Fornitore: Apollo Scientific

OR304428-250MG OR304428-500MG
APOSOR304428-250MGEA 835 EUR
APOSOR304428-250MG APOSOR304428-500MG
4-Oxa-1,9-diazaspiro[5.5]undecane dihydrochloride
4-Oxa-1,9-diazaspiro[5.5]undecane dihydrochloride
Formula: C₈H₁₆N₂O·2ClH
Peso molecolare: 229,147 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD26096846
CAS Number: 1439897-97-8

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR