Avanafil
Fornitore: Apollo Scientific
Synonyms:
4-[(3-Chloro-4-methoxybenzyl)amino]-2-[(2S)-2-(hydroxymethyl)pyrrolidin-1-yl]-N-(pyrimidin-2-ylmethyl)pyrimidine-5-carboxamide, Avanfil, (S)-4-((3-Chloro-4-methoxybenzyl)amino)-2-(2-(hydroxymethyl)pyrrolidin-1-yl)-N-(pyrimidin-2-ylmethyl)pyrimidine-5-carboxamide
Formula:
C₂₃H₂₆ClN₇O₃ Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD11973638 CAS Number: 330784-47-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...