Stampa…

Methyl 1-chloroisoquinoline-7-carboxylate

Fornitore: Apollo Scientific
Attenzione

Synonyms: 1-Chloro-7-(methoxycarbonyl)-2-azanaphthalene, 1-Chloro-7-(methoxycarbonyl)isoquinoline

OR300108-500MG OR300108-100MG
APOSOR300108-500MGEA 364 EUR
APOSOR300108-500MG APOSOR300108-100MG
Methyl 1-chloroisoquinoline-7-carboxylate
Methyl 1-chloroisoquinoline-7-carboxylate
Formula: C₁₁H₈ClNO₂
Peso molecolare: 221,64 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD18250895
CAS Number: 1206975-02-1

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR