4-Chloro-1,2-dihydro-2-oxoquinoline-3-carbonitrile
Fornitore: Apollo Scientific
|
Attenzione
|
Synonyms:
4-Chloro-2-oxo-1,2-dihydroquinoline-3-carbonitrile, 4-Chloro-3-cyano-1,2-dihydro-2-oxoquinoline
Formula:
C₁₀H₅ClN₂O Peso molecolare: 204.613 g/mol Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD01570117 CAS Number: 680210-85-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...