Stampa…

4,5,8-Trichloroquinoline

Fornitore: Apollo Scientific

OR183471-1G OR183471-250MG
APOSOR183471-1GEA 464 EUR
APOSOR183471-1G APOSOR183471-250MG
4,5,8-Trichloroquinoline
4,5,8-Trichloroquinoline
Formula: C₉H₄Cl₃N
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD09787693
CAS Number: 855763-24-5

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR